2-Bromo-5-(trifluoromethyl)pyridine ≥97% for synthesis

Supplier: Matrix Scientific

001382-25G 001382-100G
101781-536EA 56.92 USD
101781-536 101781-538
2-Bromo-5-(trifluoromethyl)pyridine ≥97% for synthesis
2-Bromo-5-(trifluoromethyl)pyridine

Hazard code(s): Irritant

Formula: C₆H₃BrF₃N
MW: 226
Boiling Pt: 78…81 °C (30mm)
Melting Pt: 45…47 °C
MDL Number: MFCD00153086
CAS Number: 50488-42-1

Order Now

Specification Test Results

Boiling point 78-81°C/30m
Melting point 45-47°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR