1-Bromo-4-(trifluoromethoxy)benzene ≥99%

Supplier: Matrix Scientific

Synonyms: 4-(Trifluoromethoxy)bromobenzene

001376-25G 001376-100G
101781-516EA 26.15 USD
101781-516 101781-518
1-Bromo-4-(trifluoromethoxy)benzene ≥99%
1-Bromo-4-(trifluoromethoxy)benzene

Hazard code(s): Irritant

Formula: C₇H₄BrF₃O
MW: 241.01 g/mol
Boiling Pt: 153…155 °C
Density: 1.62 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00040834
CAS Number: 407-14-7
EINECS: 206-979-2
UN: 3334
ADR: 9,

Order Now

Specification Test Results

Boiling point 153-155°C
Density 1.622
ND20 1.461

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR