2-Nitro-α,α,α-trifluoro-p-toluic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Nitro-4-(trifluoromethyl)benzoic acid, 2-Nitro-4-trifluoromethylbenzoic acid

001362-25G 001362-100G
101781-492EA 53.85 USD
101781-492 101781-494
2-Nitro-α,α,α-trifluoro-p-toluic acid ≥97%
2-Nitro-α,α,α-trifluoro-p-toluic acid

Hazard code(s): Irritant

Formula: C₈H₄F₃NO₄
MW: 235.12 g/mol
Melting Pt: 134…138 °C
MDL Number: MFCD00007142
CAS Number: 320-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR