5-Nitro-α,α,α-trifluoro-m-toluic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Nitro-5-(trifluoromethyl)benzoic acid
001352-5G
001352-25G
101781-464EA
29.23
USD
101781-464
101781-466
5-Nitro-α,α,α-trifluoro-m-toluic acid ≥97%
5-Nitro-α,α,α-trifluoro-m-toluic acid
Hazard code(s): Irritant
Formula:
C₈H₄F₃NO₄ MW: 235.12 g/mol Melting Pt: 128…129 °C |
MDL Number:
MFCD00024509 CAS Number: 328-80-3 |
Specification Test Results
Melting point | 128-129°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...