2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole ≥98%
Supplier: Matrix Scientific
Synonyms:
2-Amino-5-trifluoromethyl-1,3,4-thiadiazole
001339-1G
001339-5G
101781-430EA
20
USD
101781-430
101781-432
2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole ≥98%
2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole
Hazard code(s): Irritant
Formula:
C₃H₂F₃N₃S MW: 169.13 g/mol Melting Pt: 225…227 °C |
MDL Number:
MFCD00003109 CAS Number: 10444-89-0 EINECS: 233-930-2 |
Specification Test Results
Melting point | 225-227°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...