2-Amino-5-(trifluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 5-(Trifluoromethyl)pyridin-2-amine, 5-(Trifluoromethyl)-2-pyridylamine

001337-25G 001337-100G
101781-426EA 156.92 USD
101781-426 101781-428
2-Amino-5-(trifluoromethyl)pyridine ≥97%
2-Amino-5-(trifluoromethyl)pyridine

Hazard code(s): Irritant

Formula: C₆H₅F₃N₂
MW: 162.11 g/mol
Boiling Pt: 90…92 °C (20mm)
Melting Pt: 42…44 °C
MDL Number: MFCD00042164
CAS Number: 74784-70-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 90-92°C/20m
Melting point 42-44°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR