5-Amino-α,α,α-trifluoro-m-toluic acid ≥97%
Supplier: Matrix Scientific
001335-1G
001335-5G
001335-25G
101781-416EA
20
USD
101781-416
101781-418
101781-420
5-Amino-α,α,α-trifluoro-m-toluic acid ≥97%
5-Amino-α,α,α-trifluoro-m-toluic acid
Hazard code(s): Irritant
Formula:
C₈H₆F₃NO₂ MW: 205.14 g/mol |
MDL Number:
MFCD00236641 CAS Number: 328-68-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...