2-(4-Methoxyphenoxy)-5-(trifluoromethyl)aniline

Supplier: Matrix Scientific

001323-1G 001323-5G
101781-398EA 40 USD
101781-398 101781-400
2-(4-Methoxyphenoxy)-5-(trifluoromethyl)aniline
2-(4-Methoxyphenoxy)-5-(trifluoromethyl)aniline

Hazard code(s): Irritant

Formula: C₁₄H₁₂F₃NO₂
Melting Pt: 73…75 °C
MDL Number: MFCD00792432
CAS Number: 62966-74-9

Order Now

Specification Test Results

Melting point 73-75°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR