1-Bromo-2,6-difluorobenzene ≥98%

Supplier: Matrix Scientific
Danger

001313-25G 001313-100G
101781-388EA 73.85 USD
101781-388 101781-390
1-Bromo-2,6-difluorobenzene ≥98%
1-Bromo-2,6-difluorobenzene

Hazard code(s): Flammable

Formula: C₆H₃BrF₂
Boiling Pt: 61 °C (35mm)
Density: 1.71 g/cm³ (20 °C)
MDL Number: MFCD00009894
CAS Number: 64248-56-2
EINECS: 264-750-2
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 61°C/35mm
ND20 1.51


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR