2-Bromo-4,6-difluorophenol ≥97%

Supplier: Matrix Scientific
Warning

001311-5G 001311-25G 001311-100G
101781-382EA 53.85 USD
101781-382 101781-384 101781-386
2-Bromo-4,6-difluorophenol ≥97%
2-Bromo-4,6-difluorophenol

Hazard code(s): Irritant

Formula: C₆H₃BrF₂O
Boiling Pt: 153…154 °C
MDL Number: MFCD00070752
CAS Number: 98130-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR