2-Bromo-4,6-difluoroanisole ≥98%
Supplier: Matrix Scientific
|
Warning
|
001310-5G
001310-25G
101781-378EA
33.85
USD
101781-378
101781-380
2-Bromo-4,6-difluoroanisole ≥98%
2-Bromo-4,6-difluoroanisole
Hazard code(s): Irritant
Formula:
C₇H₅BrF₂O MW: 223.02 g/mol |
MDL Number:
MFCD00070754 CAS Number: 202865-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...