4-Bromo-2,6-difluoroaniline ≥98%

Supplier: Matrix Scientific

001309-25G 001309-100G
101781-374EA 30.77 USD
101781-374 101781-376
4-Bromo-2,6-difluoroaniline ≥98%
4-Bromo-2,6-difluoroaniline

Hazard code(s): Irritant

Formula: C₆H₄BrF₂N
MW: 208.01 g/mol
Melting Pt: 63…65 °C
MDL Number: MFCD00013389
CAS Number: 67567-26-4

Order Now

Specification Test Results

Melting point 63-65°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR