4-Bromo-2,6-difluorophenol ≥98%
Supplier: Matrix Scientific
|
Warning
|
001307-5G
001307-25G
101781-368EA
66.15
USD
101781-368
101781-370
4-Bromo-2,6-difluorophenol ≥98%
4-Bromo-2,6-difluorophenol
Hazard code(s): Toxic
Formula:
C₆H₃BrF₂O Melting Pt: 50 °C |
MDL Number:
MFCD00236213 CAS Number: 104197-13-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...