2-Bromo-4,6-difluoroaniline ≥97%

Supplier: Matrix Scientific
Warning

001305-25G 001305-100G
101781-364EA 33.85 USD
101781-364 101781-366
2-Bromo-4,6-difluoroaniline ≥97%
2-Bromo-4,6-difluoroaniline

Hazard code(s): Irritant

Formula: C₆H₄BrF₂N
Boiling Pt: 227 °C (1013 hPa)
Melting Pt: 42 - 42.5 °C
MDL Number: MFCD00009639
CAS Number: 444-14-4
EINECS: 429-430-0
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Melting point 41-42°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR