4-Bromo-2,6-difluoroanisole ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 5-Bromo-1,3-difluoro-2-methoxybenzene

001304-1G 001304-5G
101781-360EA 29.23 USD
101781-360 101781-362
4-Bromo-2,6-difluoroanisole ≥97%
4-Bromo-2,6-difluoroanisole

Hazard code(s): Irritant

Formula: C₇H₅BrF₂O
MW: 223.02 g/mol
Boiling Pt: 100 °C (15mm)
MDL Number: MFCD01631351
CAS Number: 104197-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR