2-Bromo-1,4-dimethoxybenzene ≥98%

Supplier: Matrix Scientific
Warning

001292-25G 001292-100G
101781-336EA 64.62 USD
101781-336 101781-338
2-Bromo-1,4-dimethoxybenzene ≥98%
2-Bromo-1,4-dimethoxybenzene

Hazard code(s): Irritant

Formula: C₈H₉BrO₂
Boiling Pt: 130…131 °C (10mm)
Density: 1.45 g/cm³
MDL Number: MFCD00008355
CAS Number: 25245-34-5
EINECS: 246-756-7

Order Now

Specification Test Results

Boiling point 130-131°C/1
Density 1.45
ND20 1.57


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR