2-Bromo-4,5-dimethoxybenzoic acid ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 6-Bromoveratric acid

001289-5G 001289-25G
101781-324EA 30.77 USD
101781-324 101781-326
2-Bromo-4,5-dimethoxybenzoic acid ≥98%
2-Bromo-4,5-dimethoxybenzoic acid

Hazard code(s): Irritant

Formula: C₉H₉BrO₄
MW: 261.07 g/mol
Melting Pt: 190…193 °C
MDL Number: MFCD00017542
CAS Number: 6286-46-0

Order Now

Specification Test Results

Melting point 190-193°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR