2-Bromo-4,5-difluoroanisole ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 1-Bromo-4,5-difluoro-2-methoxybenzene

001286-5G 001286-25G 001286-100G
101781-318EA 44.62 USD
101781-318 101781-320 101781-322
2-Bromo-4,5-difluoroanisole ≥98%
2-Bromo-4,5-difluoroanisole

Hazard code(s): Irritant

Formula: C₇H₅BrF₂O
MW: 223.02 g/mol
Melting Pt: 23…25 °C
MDL Number: MFCD00070753
CAS Number: 202865-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR