4-Bromo-3,5-difluoroaniline ≥98%

Supplier: Matrix Scientific

001285-25G 001285-100G
101781-314EA 224.62 USD
101781-314 101781-316
4-Bromo-3,5-difluoroaniline ≥98%
4-Bromo-3,5-difluoroaniline

Hazard code(s): Toxic

Formula: C₆H₄BrF₂N
MW: 208.01 g/mol
Melting Pt: 99…101 °C
MDL Number: MFCD00236593
CAS Number: 203302-95-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR