1-Bromo-2,5-difluoro-4-nitrobenzene ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Brom-2,5-difluor-4-nitrobenzol, 4-Bromo-2,5-difluoronitrobenzene

001282-5G 001282-25G
101781-304EA 29.23 USD
101781-304 101781-306
1-Bromo-2,5-difluoro-4-nitrobenzene ≥97%
1-Bromo-2,5-difluoro-4-nitrobenzene

Hazard code(s): Irritant

Formula: C₆H₂BrF₂NO₂
MW: 237.99 g/mol
Melting Pt: 55 - 57 °C
MDL Number: MFCD00042184
CAS Number: 167415-27-2

Order Now

Specification Test Results

Melting point 55-57°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR