4-Bromo-2,5-difluoroaniline ≥97%

Supplier: Matrix Scientific

001280-25G 001280-100G
101781-300EA 70.77 USD
101781-300 102550-792
4-Bromo-2,5-difluoroaniline ≥97%
4-Bromo-2,5-difluoroaniline

Hazard code(s): Irritant

Formula: C₆H₄BrF₂N
Melting Pt: 74…75 °C
MDL Number: MFCD01861125
CAS Number: 112279-60-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 74-75°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR