2-Bromo-4,5-difluorophenol ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 1-Bromo-3,4-difluoro-6-hydroxybenzene

001278-5G 001278-25G
101781-294EA 32.31 USD
101781-294 101781-296
2-Bromo-4,5-difluorophenol ≥97%
2-Bromo-4,5-difluorophenol

Hazard code(s): Irritant

Formula: C₆H₃BrF₂O
MW: 208.99 g/mol
Density: 1.829 g/cm³
MDL Number: MFCD00070751
CAS Number: 166281-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR