2-Bromo-1,4-dichlorobenzene ≥98% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 1-Bromo-2,5-dichlorobenzene

001273-25G 001273-100G
101781-284EA 66.15 USD
101781-284 101781-286
2-Bromo-1,4-dichlorobenzene ≥98% for synthesis
2-Bromo-1,4-dichlorobenzene

Hazard code(s): Irritant

Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
Boiling Pt: 235 °C
Melting Pt: 34…36 °C
MDL Number: MFCD00018505
CAS Number: 1435-50-3
EINECS: 215-859-9

Order Now

Specification Test Results

Boiling point 235°C
Melting point 34-36°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR