1-Bromo-2,5-dichloro-3-fluorobenzene ≥98%
Supplier: Matrix Scientific
|
Warning
|
001272-5G
001272-25G
101781-280EA
35.38
USD
101781-280
101781-282
1-Bromo-2,5-dichloro-3-fluorobenzene ≥98%
1-Bromo-2,5-dichloro-3-fluorobenzene
Hazard code(s): Irritant
Formula:
C₆H₂BrCl₂F MW: 243.89 g/mol |
MDL Number:
MFCD00142585 CAS Number: 202865-57-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...