2-Amino-3,5-dibromo-6-methylpyridine ≥97% for synthesis
Supplier: Matrix Scientific
Synonyms:
3,5-Dibromo-6-methyl-2-pyridinamine
001271-5G
001271-25G
101781-276EA
15.38
USD
101781-276
101781-278
2-Amino-3,5-dibromo-6-methylpyridine ≥97% for synthesis
2-Amino-3,5-dibromo-6-methylpyridine
Hazard code(s): Irritant
Formula:
C₆H₆Br₂N₂ Melting Pt: 144…145 °C |
MDL Number:
MFCD00068229 CAS Number: 91872-10-5 |
Specification Test Results
Melting point | 144-145°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...