3,3,4,5,5,5-Hexafluoro-2-pentanol ≥97%
Supplier: Matrix Scientific
001267-5G
001267-25G
101781-272EA
200
USD
101781-272
101781-274
3,3,4,5,5,5-Hexafluoro-2-pentanol ≥97%
3,3,4,5,5,5-Hexafluoro-2-pentanol
Hazard code(s): Irritant
Formula:
C₅H₆F₆O Boiling Pt: 119…121 °C (747mm) |
MDL Number:
MFCD00155852 CAS Number: 2711-81-1 |
Specification Test Results
Boiling point | 119-121°C/7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...