4-Bromoveratrole ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Bromo-3,4-dimethoxybenzene, 4-Bromo-1,2-dimethoxybenzene

001249-25G 001249-100G
101781-248EA 32.31 USD
101781-248 101781-250
4-Bromoveratrole ≥97%
4-Bromoveratrole

Hazard code(s): Irritant

Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Boiling Pt: 255 °C (1013 hPa)
Density: 1.5068 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008381
CAS Number: 2859-78-1
EINECS: 220-677-8

Order Now

Specification Test Results

Boiling point 124-126°C/9
Density 1.702
ND20 1.573

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR