4-Bromo-1,2-difluorobenzene ≥98%
Supplier: Matrix Scientific
001247-100G
001247-500G
101781-240EA
32.31
USD
101781-240
101781-242
4-Bromo-1,2-difluorobenzene ≥98%
4-Bromo-1,2-difluorobenzene
Hazard code(s): Irritant
Formula:
C₆H₃BrF₂ Boiling Pt: 150…151 °C Melting Pt: –4 °C Density: 1.707 g/cm³ Flash Pt: 33 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000304 CAS Number: 348-61-8 EINECS: 206-481-5 UN: 1993 ADR: 3,III |
Specification Test Results
Boiling point | 150-151°C |
Density | 1.707 |
ND20 | 1.5048 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...