4-Bromo-1,2-dichlorobenzene ≥99%

Supplier: Matrix Scientific
Warning

Synonyms: 1-Bromo-3,4-dichlorobenzene

001241-25G 001241-100G
101781-228EA 16.92 USD
101781-228 101781-230
4-Bromo-1,2-dichlorobenzene ≥99%
4-Bromo-1,2-dichlorobenzene

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: 24.5 °C
Density: 1.781 g/cm³ (20 °C)
MDL Number: MFCD00040849
CAS Number: 18282-59-2
EINECS: 242-160-6

Order Now

Specification Test Results

Boiling point 140-142°C/3
Melting point 24-25°C
Density 1.73
ND20 1.5989

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR