1-Bromo-2,4-dichlorobenzene ≥99%

Supplier: Matrix Scientific
Warning

Synonyms: 2,4-Dichlorobromobenzene

001240-5G 001240-25G
101781-224EA 21.54 USD
101781-224 101781-226
1-Bromo-2,4-dichlorobenzene ≥99%
1-Bromo-2,4-dichlorobenzene

Hazard code(s): Irritant

Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
Boiling Pt: 83 °C (4mm)
Melting Pt: 26…27 °C
MDL Number: MFCD00018333
CAS Number: 1193-72-2
EINECS: 214-778-6

Order Now

Specification Test Results

Boiling point 83°C/4mm
Melting point 26-27°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR