1-Bromo-2,4,5-trifluorobenzene ≥99%

Supplier: Matrix Scientific
Warning

Synonyms: 2,4,5-Trifluorobromobenzene

001224-25G 001224-100G
101781-200EA 41.54 USD
101781-200 101781-202
1-Bromo-2,4,5-trifluorobenzene ≥99%
1-Bromo-2,4,5-trifluorobenzene

Hazard code(s): Irritant

Formula: C₆H₂BrF₃
MW: 210.98 g/mol
Boiling Pt: 144 °C (1013 hPa)
Melting Pt: -19 °C
Density: 1.8022 g/cm³ (20 °C)
MDL Number: MFCD00000306
CAS Number: 327-52-6
EINECS: 206-318-8
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 144°C
Melting point -19°C
Density 1.802
ND20 1.486

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR