2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol ≥98%

Supplier: Matrix Scientific

Synonyms: 1H,1H,6H,6H-Perfluoro-1,6-hexanediol, 1H,1H,6H,6H-Octafluoro-1,6-hexanediol

001221-5G 001221-25G 001221-100G
101781-194EA 35.38 USD
101781-194 101781-196 101781-198
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol ≥98%
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol

Hazard code(s): Irritant

Formula: C₆H₆F₈O₂
MW: 262.1 g/mol
Boiling Pt: 100 °C (3mm)
Melting Pt: 67…70 °C
MDL Number: MFCD00069087
CAS Number: 355-74-8

Order Now

Specification Test Results

Boiling point 100°C/3mm
Melting point 67-70°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR