5-Bromo-2,3-difluorophenol ≥97%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
1-Bromo-3,4-difluoro-5-hydroxybenzene
001203-25G
001203-100G
101781-144EA
56.92
USD
101781-144
101781-146
5-Bromo-2,3-difluorophenol ≥97%
5-Bromo-2,3-difluorophenol
Hazard code(s): Irritant
Formula:
C₆H₃BrF₂O MW: 208.99 g/mol Density: 1.8 g/cm³ |
MDL Number:
MFCD01631350 CAS Number: 186590-26-1 |
Specification Test Results
ND20 | 1.538 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...