5-Bromo-2,3-difluoroanisole ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 5-Bromo-1,2-difluoro-3-methoxybenzene

001202-5G 001202-25G
101781-140EA 32.31 USD
101781-140 101781-142
5-Bromo-2,3-difluoroanisole ≥97%
5-Bromo-2,3-difluoroanisole

Hazard code(s): Irritant

Formula: C₇H₅BrF₂O
MW: 223.02 g/mol
Melting Pt: 33…36 °C
MDL Number: MFCD01631352
CAS Number: 261762-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR