1-Bromo-2,3-difluorobenzene ≥98%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
2,3-Difluorobromobenzene
001200-25G
001200-100G
101781-136EA
73.85
USD
101781-136
101781-138
1-Bromo-2,3-difluorobenzene ≥98%
1-Bromo-2,3-difluorobenzene
Hazard code(s): Irritant
Formula:
C₆H₃BrF₂ MW: 192.99 g/mol Boiling Pt: 69…70 °C (12mm) Density: 1.72 g/cm³ (20 °C) |
MDL Number:
MFCD00061136 CAS Number: 38573-88-5 |
Specification Test Results
Boiling point | 69-70°C/12m |
Density | 1.724 |
ND20 | 1.509 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...