1-Bromo-2,3,5-trifluorobenzene ≥95%

Supplier: Matrix Scientific
Danger

Synonyms: 2,3,5-Trifluorobromobenzene

001182-25G 001182-100G
101781-104EA 73.85 USD
101781-104 101781-106
1-Bromo-2,3,5-trifluorobenzene ≥95%
1-Bromo-2,3,5-trifluorobenzene

Hazard code(s): Irritant

Formula: C₆H₂BrF₃
MW: 210.98 g/mol
Density: 1.758 g/cm³
MDL Number: MFCD00012232
CAS Number: 133739-70-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Density 1.758
ND20 1.486

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR