2-Bromo-3,3,3-trifluoropropene ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-3,3,3-trifluoro-1-propene

001168-25G 001168-100G
101781-066EA 107.69 USD
101781-066 101781-068
2-Bromo-3,3,3-trifluoropropene ≥98%
2-Bromo-3,3,3-trifluoropropene

Hazard code(s): Flammable, Refrigerate

Formula: C₃H₂BrF₃
MW: 174.95 g/mol
Boiling Pt: 34…35 °C
Density: 1.686 g/cm³
MDL Number: MFCD00077469
CAS Number: 1514-82-5

Order Now

Specification Test Results

Boiling point 34-35°C
Density 1.686
ND20 1.3523

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR