Methyl-3-(trifluoromethyl)benzoate ≥98%

Supplier: Matrix Scientific

Synonyms: Methyl 3-(trifluoromethyl)benzoate

001111-5G 001111-25G
101780-988EA 15.38 USD
101780-988 101780-990
Methyl-3-(trifluoromethyl)benzoate ≥98%
Methyl-3-(trifluoromethyl)benzoate

Hazard code(s): Irritant

Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
Boiling Pt: 80…82 °C (15mm)
Density: 1.295 g/cm³
MDL Number: MFCD00043543
CAS Number: 2557-13-3

Order Now

Specification Test Results

Boiling point 80-82°C/15m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR