Methyl-2-(trifluoromethyl)benzoate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-(trifluoromethyl)benzoate

001105-5G 001105-25G
101780-970EA 27.69 USD
101780-970 101780-972
Methyl-2-(trifluoromethyl)benzoate ≥97%
Methyl-2-(trifluoromethyl)benzoate

Hazard code(s): Irritant

Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
Boiling Pt: 104…105 °C (25mm)
Density: 1.306 g/cm³
MDL Number: MFCD00236050
CAS Number: 344-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR