3',5'-Bis(trifluoromethyl)acetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 1,3-Di(trifluormethyl)-5-ethanoylbenzol, 1-(3,5-Bis(trifluoromethyl)phenyl)ethanone, 1-[3,5-Bis(trifluoromethyl)phenyl]ethan-1-one

001101-25G 001101-100G
101780-958EA 36.92 USD
101780-958 101780-960
3',5'-Bis(trifluoromethyl)acetophenone ≥97%
3',5'-Bis(trifluoromethyl)acetophenone

Hazard code(s): Irritant

Formula: C₁₀H₆F₆O
MW: 256.15 g/mol
Boiling Pt: 85 °C (8mm)
Density: 1.42 g/cm³ (20 °C)
MDL Number: MFCD00009910
CAS Number: 30071-93-3
EINECS: 250-023-7

Order Now

Specification Test Results

Density 1.422
ND20 1.4221

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR