Nonafluoro(trifluoromethyl)cyclopentane ≥90%

Supplier: Matrix Scientific

001094-25G 001094-100G
101780-936EA 44.62 USD
101780-936 101780-938
Nonafluoro(trifluoromethyl)cyclopentane ≥90%
Nonafluoro(trifluoromethyl)cyclopentane

Hazard code(s): Irritant

Formula: C₆F₁₂
MW: 300.05 g/mol
Boiling Pt: 48 °C
MDL Number: MFCD00066611
CAS Number: 1805-22-7
EINECS: 217-298-5

Order Now

Specification Test Results

Boiling point 48°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR