Potassium hydroxide 0.0500 Normal (N/20), Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility
Supplier: Ricca Chemical
Synonyms:
Potash caustic, Caustic potash
R6198000-500A
R6198000-1A
RC6198-16EA
57
USD
RC6198-16
RC6198-32
Potassium hydroxide 0.0500 Normal (N/20), Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility
Potassium hydroxide
Formula:
KOH MW: 56.11 g/mol Boiling Pt: 1320 Melting Pt: 360 Storage Temperature: Ambient |
MDL Number:
MFCD00003553 CAS Number: 1310-58-3 EINECS: 215-181-3 UN: 1814 ADR: 8 II Merck Index: 12,07806 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 84) | 0.05000 ± 0.00005 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...