Sodium hydroxide 0.333 Normal (N/3), Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility

Supplier: Ricca Chemical
Warning

Synonyms: Caustic soda

R7377000-20F R7377000-1A R7377000-4A
RC7377-5EA 289 USD
RC7377-5 RC7377-32 RC7377-1
Sodium hydroxide 0.333 Normal (N/3), Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 1.02 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 III
Merck Index: 14,08627

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 0.3330 ± 0.0001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR