Sulfuric acid 18.0 N

Supplier: Ricca Chemical
Danger

R8343180-500A R8343180-1A R8343180-2.5D R8343180-4A
RC8343.18-16EA 105 USD
RC8343.18-16 RC8343.18-32 RC8343.18-5PT RCR8343180-4A
Sulfuric acid 18.0 N
Sulfuric acid
Formula: H₂SO₄
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 17.95-18.05 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR