Sodium hydroxide 0,400 ± 0,004% (w/v) in aqueous solution analytical reagent
Supplier: Ricca Chemical
Synonyms:
Caustic soda
R7242500-1A
R7242500-500A
RC7242.5-32EA
41
USD
RC7242.5-32
RC7242.5-16
Sodium hydroxide 0,400 ± 0,004% (w/v) in aqueous solution analytical reagent
Sodium hydroxide
Formula:
NaOH MW: 40 g/mol Boiling Pt: 1390 Melting Pt: 323 Density: 1.003 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8 III Merck Index: 14,08627 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 723) | 0.400 ± 0.004 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...