Acetone:Alcohol (1:1) for decolorizing, for Gram staining
Supplier: Ricca Chemical
|
Danger
|
R0200000-4A
R0200000-500A
R0200000-1A
RC2001EA
352
USD
RC2001
RC20016
RC20032
Acetone:Alcohol (1:1) for decolorizing, for Gram staining
Acetone:Alcohol
Acetone-Alcohol, 1 + 1 Decolorizer Solution for use in Gram Staining
Flash Pt:
–18 °C Storage Temperature: Ambient |
ADR:
3,II |
Specification Test Results
Appearance | Clear, colorless, alcohol odor |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...