Potassium hydroxide 1.00 N in denatured alcohol
Supplier: Ricca Chemical
|
Danger
|
R6266900-4A
R6266900-1A
RC6266.9-1EA
520
USD
RC6266.9-1
RC6266.9-32
Potassium hydroxide 1.00 N in denatured alcohol
Potassium hydroxide
Formula:
KOH Storage Temperature: Ambient |
MDL Number:
MFCD00003553 CAS Number: 1310-58-3 EINECS: 215-181-3 UN: 2924 ADR: 3,II |
Specification Test Results
Appearance | Clear, colorless, alcohol odor |
Assay at 20 °C (traceable to NIST SRM 723) | 1.000 ± 0.005 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...