Potassium hydroxide 0.500 N (N/2) in denatured alcohol

Supplier: Ricca Chemical
Danger

R6265000-4A R6265000-1A R6265000-20F
RC62651EA 378 USD
RC62651 RC6265-32 RCR6265000-20F
Potassium hydroxide 0.500 N (N/2) in denatured alcohol
Potassium hydroxide
Formula: KOH
Melting Pt: Data not available.
Density: 0.82
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 2924
ADR: 3 (8),II

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 0.4995-0.5005 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR