Potassium hydroxide 0.250 Normal (N/4), Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility

Supplier: Ricca Chemical
Warning

Synonyms: Potash caustic, Caustic potash

R6220000-1A R6220000-4A R6220000-20F
RC6220-32EA 96 USD
RC6220-32 RC6220-1 RC6220-5
Potassium hydroxide 0.250 Normal (N/4), Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8 II
Merck Index: 12,07806

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 0.2500 ± 0.0001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR