Iodine (iodine-iodide) 0.0100 N (N/100) in aqueous solution
Supplier: Ricca Chemical
R3971000-4C
R3971000-1C
R3971000-500C
RC3971-1EA
322
USD
RC3971-1
RC3971-32
RC3971-16
Iodine (iodine-iodide) 0.0100 N (N/100) in aqueous solution
Iodine (iodine-iodide)
Storage Temperature:
Ambient |
MDL Number:
MFCD00011355 EINECS: 231-442-4 UN: 3264 ADR: 8,II |
Specification Test Results
Appearance | Clear, brown, Iodine odor |
Assay at 20 °C (traceable to NIST SRM 136) | 0.01000 ± 0.00002 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...